Learn More
ICP-MS Pharma & USP 232 Internal Standard Solution: Bi, Ga, In @ 100 ug/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
ICP-MS Pharma & USP 232 Internal Standard Solution: Bi, Ga, In @ 100 ug/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Bismuth/Gallium/Indium |
| CAS | 7440-69-9, 7440-55-3, 7440-74-6 |
| Analyte or Component Names | Bismuth/Gallium/Indium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Bismuth 100 μg/mL/Gallium 100 μg/mL/Indium 100 μg/mL |