missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Naval Brass, High Leaded, CDA 485, C48500, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Naval Brass, High Leaded, CDA 485, C48500, 100g
Specifications
Specifications
| Chemical Name or Material | Cadmium/Manganese/Antimony/Aluminium/Carbon/Silicon/Nickel/Arsenic/Iron/Tin/Lead/Zinc/Copper/Chromium/Hafnium/Hydrogen/Palladium/Tantalum/Tungsten/Yttrium/Beryllium/Boron/Calcium/Molybdenum/Niobium/Vanadium/Zirconium/Magnesium/Cobalt/Bismuth/Nitrogen/Titanium/Sulfur/Oxygen/Phosphorus/Selenium/Silver |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0005 %/Manganese 0.0013 %/Antimony 0.0018 %/Aluminium 0.002 %/Carbon 0.002 %/Silicon 0.003 %/Nickel 0.013 %/Arsenic 0.055 %/Iron 0.062 %/Tin 0.759 %/Lead 1.76 %/Zinc 36.5 %/Copper 60.8 %/Chromium (<0.0010) %/Hafnium (<0.0010) %/Hydrogen (<0.0010) %/Palladium (<0.0010) %/Tantalum (<0.0010) %/Tungsten (<0.0010) %/Yttrium (<0.0010) %/Beryllium (<0.0050) %/Boron (<0.0050) %/Calcium (<0.0050) %/Molybdenum (<0.0050) %/Niobium (<0.0050) %/Vanadium (<0.0050) %/Zirconium (<0.0050) %/Magnesium (0.0001) %/Cobalt (0.0005) %/Bismuth (0.001) %/Nitrogen (0.001) %/Titanium (0.001) %/Sulfur (0.0019) %/Oxygen (0.002) %/Phosphorus (0.002) %/Selenium (0.003) %/Silver (0.0031) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction