Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Minimizes the hands-on time involved in extraction. Macherey Nagel™ Cellulose Extraction Thimbles are used for holding solid materials from which substances are to be extracted with a suitable solvent.
Brand:  Macherey-Nagel™ 645020
Product Code. 10092721
  • ONLINE EXCLUSIVE PRICE
    97.90 EUR
    94.80 EUR / Pack of 25
    Save 3.10 EUR
    3% Off
Description
  • Manufactured from pure cellulose
  • Used in the fields of air and waste gas analysis for collecting solid particles
  • The choice of analytical chemists using Soxhlet or automated extractors
Specifications
Specifications
25 Pk.
Cellulose
80 mm
30 mm
Analysis of soil contaminants, Analysis of soil contaminants
Extraction Thimbles
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions