missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 6 Mda Fluorescein Hyaluronic Acid

Product Code. 30216522
Encompass_Preferred
Click to view available options
:
100 mg
This item is not returnable. View return policy

Product Code. 30216522

Brand: TdB Labs AB FHASe_100mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescein-hyaluronic acid 6 MDa is a high molecular weight, green-fluorescent biopolymer used in diffusion, retention, and matrix interaction studies. Ideal for tracing HA behavior in tissue models and drug delivery systems.

Product Overview: Fluorescein-hyaluronic acid 6 MDa is a high molecular weight hyaluronic acid conjugated with fluorescein, providing strong green fluorescence for biological imaging, molecular tracking, and barrier function studies. With an approximate molecular weight of 6 million Daltons, this product is designed for modeling the behavior of extremely large biomolecules and polymers in biological environments.

  • Features:
    • High Molecular Weight: Approximately 6 MDa, offering properties similar to naturally occurring extracellular matrix components.
    • Green Fluorescence: Excitation maximum at 495 nm and emission maximum at 519 nm, compatible with standard FITC filter settings.
    • Water-Soluble: Readily dissolves in aqueous solutions, facilitating easy preparation for biological studies.
    • Biocompatible Backbone: Hyaluronic acid is naturally occurring and well-tolerated in biological systems, supporting use in live cell and tissue models.
    • Controlled Degree of Labeling: Ensures bright fluorescence while preserving native polymer characteristics and behavior.
  • Benefits:
    • Ideal for ECM and Barrier Studies: Models large molecule transport in extracellular matrix environments and across biological barriers.
    • Visualizes Molecular Movement: Tracks the diffusion and distribution of high molecular weight biomolecules in tissue and hydrogel systems.
    • Suitable for Live and Fixed Imaging: Can be used in both dynamic live-cell setups and after fixation for endpoint assays.
    • Represents Native Biological Behavior: High molecular weight closely mimics natural hyaluronic acid found in tissues, enabling physiologically relevant studies.
    • Stable, Bright Fluorescence: Provides consistent, high-intensity signal for reliable imaging and quantitative analysis.
  • Applications:
    • Extracellular Matrix Research: Used to model diffusion and retention of large biomolecules in native and engineered tissue systems.
    • Barrier Permeability Studies: Tracks the ability of macromolecules to cross biological barriers like endothelial and epithelial layers.
    • Tissue Engineering and Regenerative Medicine: Studies integration and movement of large molecules in scaffold-based and hydrogel-based systems.
    • Drug Delivery Research: Models nanoparticle or macromolecular therapeutic diffusion through tissues.
    • In Vivo and In Vitro Imaging: Used to visualize macromolecule distribution in animal models and cultured tissues.
  • Technical Specifications:
    • Fluorescence: Excitation maximum at 495 nm, emission maximum at 519 nm.
    • Molecular Weight: Approximately 6 million Daltons (6 MDa).
    • Appearance: Light-sensitive, greenish dry powder, fully water-soluble.
    • Labeling: Fluorescein conjugation optimized for strong fluorescence with minimal alteration of HA's biological properties.
  • Storage and Stability:
    • Store dry and protected from light at 2°C to 8°C for optimal stability.
    • Stable for extended periods when stored under recommended conditions.
  • Packaging and Availability:
    • Supplied as dry powder for ease of storage and handling.
    • Available in various research packaging sizes to support different experimental needs.

Specifikationer

Detection Method Fluorescence
Format Bottle
Product Type Reagent
Quantity 100 mg
Form Powder
Korrigering av produktinnehåll

Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.