missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Wnt-2/sFRP-1 Complex Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Measured by its ability to activate Wnt induced TCF reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 10.0-150 ng/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS and NaCl with Trehalose. |
| Gene ID (Entrez) | 7472 |
| Molecular Weight (g/mol) | M.W.-observed: 30-45 kDa, under reducing conditions., M.W.-theroretical: 38 kDa (Wnt-2) & 33 kDa (sFRP-1) |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Wnt-2 protein Human Wnt-2 (Ser26-Thr360) (C-terminus) Accession # P09544.1 Human sFRP-1 (Ser32-Lys314) (C-terminus) Accession # AAB70793.1 (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | FrzA, INT1L1, INT1L1secreted growth factor, Int-1-like protein 1, Int-1-related protein, IRP, IRPprotein Wnt-2, SARP-2, sFRP1, wingless-type MMTV integration site family member 2, Wnt2 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction