Learn More
10-Phenyl-1-decanol, 96%, Thermo Scientific™
Product Code. 11338335
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11338335 5 g 5g
11328335 1 g 1g
2 options
Product Code. 11338335
Brand: Thermo Scientific Alfa Aesar L10269.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62607-69-6
Molecular Formula C16H26O
Molecular Weight (g/mol) 234.383
MDL Number MFCD00014426
InChI Key WNFXODBOMMSELY-UHFFFAOYSA-N
Synonym 10-phenyl-1-decanol, benzenedecanol, acmc-20aozo, 10-phenyl-n-decanol, 10-phenyl-1-decyl alcohol
PubChem CID 568845
IUPAC Name 10-phenyldecan-1-ol
SMILES C1=CC=C(C=C1)CCCCCCCCCCO
Specifications
Melting Point 35°C to 37°C
Boiling Point 145°C to 147°C (0.5 mmHg)
Quantity 5 g
Beilstein 1955067
Formula Weight 234.39
Percent Purity 96%
Chemical Name or Material 10-Phenyl-1-decanol