Learn More
10-Azido-1-decanol, TRC
Specifications
Specifications
| Chemical Name or Material | 10-Azido-1-decanol |
| CAS | 57395-48-9 |
| Molecular Formula | C10 H21 N3 O |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H21N3O/c11-13-12-9-7-5-3-1-2-4-6-8-10-14/h14H,1-10H2 |
| SMILES | OCCCCCCCCCCN=[N+]=[N-] |
| IUPAC Name | 10-azidodecan-1-ol |
| Molecular Weight (g/mol) | 199.29 |
| Formula Weight | 199.1685 |