Learn More
Nalgene Environmental 1 x 144 -40mL Amber Vial,
GREENER_CHOICE
Brand:  Nalgene Environmental 141-40A/DB
1 x 144 -40mL Amber Vial,
Product Code. 13595110
  • ONLINE EXCLUSIVE PRICE
    357.00 EUR
    342.01 EUR / Each
    Save 14.99 EUR
    4% Off
Only null left
Estimated Shipment: 29-12-2025