Learn More
1-Trimethylsilyl-1,2,4-triazole, 95%
Product Code. 11444567
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444567 25 g 25g
11434567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11444567 Supplier Thermo Scientific Alfa Aesar Supplier No. B21105.14
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
CAS: 18293-54-4 | C5H11N3Si | 141.25 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18293-54-4
Molecular Formula C5H11N3Si
Molecular Weight (g/mol) 141.25
MDL Number MFCD00046070
InChI Key WPSPBNRWECRRPK-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1,2,4-triazole, 1-trimethylsilyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 1-trimethylsilyl, n-trimethylsilyl-1,2,4-triazole, acmc-20aorq, 1-trimethylsilyl-1,2,4 triazole, 1h-1,2,4-triazole, tms derivative, trimethyl 1,2,4-triazol-1-yl silane, 1h-1,2,4-triazole,1-trimethylsilyl
PubChem CID 87558
IUPAC Name trimethyl(1,2,4-triazol-1-yl)silane
SMILES C[Si](C)(C)N1C=NC=N1
Specifications
Density 0.98
Boiling Point 73°C to 75°C (12 mmHg)
Flash Point −5°C (23°F)
Refractive Index 1.461
Quantity 25 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 141.25
Percent Purity 98%
Chemical Name or Material 1-Trimethylsilyl-1,2,4-triazole