Learn More
1-Trimethylsilyl-1,2,4-triazole, 95%
Product Code. 11434567
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11434567
Brand: Thermo Scientific Alfa Aesar B21105.06
Request Bulk or Custom Format
CAS: 18293-54-4 | C5H11N3Si | 141.25 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18293-54-4
Molecular Formula C5H11N3Si
Molecular Weight (g/mol) 141.25
MDL Number MFCD00046070
InChI Key WPSPBNRWECRRPK-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1,2,4-triazole, 1-trimethylsilyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 1-trimethylsilyl, n-trimethylsilyl-1,2,4-triazole, acmc-20aorq, 1-trimethylsilyl-1,2,4 triazole, 1h-1,2,4-triazole, tms derivative, trimethyl 1,2,4-triazol-1-yl silane, 1h-1,2,4-triazole,1-trimethylsilyl
PubChem CID 87558
IUPAC Name trimethyl(1,2,4-triazol-1-yl)silane
SMILES C[Si](C)(C)N1C=NC=N1
Specifications
Density 0.98
Boiling Point 73°C to 75°C (12 mmHg)
Flash Point −5°C (23°F)
Refractive Index 1.461
Quantity 5 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 141.25
Percent Purity 98%
Chemical Name or Material 1-Trimethylsilyl-1,2,4-triazole