missing translation for 'onlineSavingsMsg'
Learn More

1-Thio-beta-D-glucose tetraacetate, 98+%

Product Code. p-7045628
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357756 250 mg 250mg
11347756 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357756 Supplier Thermo Scientific Alfa Aesar Supplier No. L11404.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19879-84-6 | C14H20O9S | 364.37 g/mol

1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.

Solubility
Soluble in water and chloroform (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19879-84-6
Molecular Formula C14H20O9S
Molecular Weight (g/mol) 364.37
MDL Number MFCD00063271
InChI Key SFOZKJGZNOBSHF-UHFFFAOYNA-N
Synonym 1-thio-beta-d-glucose tetraacetate, 2r,3r,4s,5r,6s-2-acetoxymethyl-6-mercaptotetrahydro-2h-pyran-3,4,5-triyl triacetate, 1-thio-b-d-glucose tetraacetate, 1-thio-beta-d-glucose 2,3,4,6-tetraacetate, 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranose, 1-thio-, a-d-glucose tetraacetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-sulfanyloxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-1-thio-, a-d-glucopyranose, beta-d-thioglucose tetraacetate, 1-thio-ss-d-glucose tetraacetate
PubChem CID 88293
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-sulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 115°C to 117°C
Quantity 250 mg
Beilstein 1440689
Solubility Information Soluble in water and chloroform (50mg/ml).
Optical Rotation +5.5° ± 1° (c=2, Chloroform)
Formula Weight 364.37
Percent Purity 99%
Chemical Name or Material 1-Thio-beta-D-glucose tetraacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.