Learn More
Microspec™ 1 Roll Autoclave Tape
Product Code. p-7085775
Click to view available options
Dimensions (L x W):
12 mm x 50 m
19 mm x 51 m
Unit Size:
Each
Product Code. 16605492
Brand: Microspec™ X3
Edge
Autoclavable tape roll
  • Autoclavable
  • Different sizes available
Specifications
Autoclavable Autoclavable
Dimensions (L x W) 19 mm x 51 m
Temperature (Metric) 121°C
Quantity 1