Learn More
1-Pyridin-3-yl-piperazine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Pyridin-3-yl-piperazine Hydrochloride |
| CAS | 1010133-97-7 |
| Molecular Formula | C9H13N3•HCl |
| Quantity | 1 g |
| Synonym | 1-(3-Pyridinyl)piperazine Hydrochloride, 1-(3-Pyridyl)piperazine Hydrochloride, 1-(Pyridin-3-yl)piperazine Hydrochloride, 3-(Piperazin-4-yl)pyridine Hydrochloride, NSC 505538 Hydrochloride |
| InChI Formula | InChI=1S/C9H13N3.ClH/c1-2-9(8-11-3-1)12-6-4-10-5-7-12;/h1-3,8,10H,4-7H2;1H |
| SMILES | C1=CC(=CN=C1)N2CCNCC2.Cl |
| IUPAC Name | 1-(pyridin-3-yl)piperazine hydrochloride |
| Molecular Weight (g/mol) | 199.68 |
| Formula Weight | 199.09 |