Learn More
1-Pyridin-2-yl-propan-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Pyridin-2-yl-propan-2-one |
| CAS | 6302-02-9 |
| Molecular Formula | C8H9NO |
| Quantity | 50 mg |
| Synonym | 1-(2-Pyridinyl)-2-propanone, (2-Pyridyl)acetone, 1-(2-Pyridyl)acetone, 1-(Pyridin-2-yl)acetone, 1-Pyridyl-2-propanone, 2-(2-Oxopropan-1-yl)pyridine, 2-Acetonylpyridine, 2-Picolyl Methyl Ketone, NSC 42754 |
| InChI Formula | InChI=1S/C8H9NO/c1-7(10)6-8-4-2-3-5-9-8/h2-5H,6H2,1H3 |
| SMILES | CC(=O)Cc1ccccn1 |
| IUPAC Name | 1-pyridin-2-ylpropan-2-one |
| Molecular Weight (g/mol) | 135.17 |
| Formula Weight | 135.0684 |