missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-propanol, 98%

Product Code. p-3211417 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10777973

Brand: Thermo Scientific Acros 183060050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14898-87-4 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-87-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004540
InChI Key WYTRYIUQUDTGSX-UHFFFAOYNA-N
Synonym 1-phenyl-2-propanol, benzyl methyl carbinol, 2-hydroxy-1-phenylpropane, benzylmethylcarbinol, 2-propanol, 1-phenyl, benzeneethanol, .alpha.-methyl, alpha-methyl-phenethyl alcohol, benzeneethanol, alpha-methyl, alpha-methylbenzeneethanol, .alpha.-methylphenethyl alcohol
PubChem CID 94185
IUPAC Name 1-phenylpropan-2-ol
SMILES CC(O)CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9900g/mL
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH(OH)CH3
Refractive Index 1.5200 to 1.522
Quantity 5 g
Beilstein 06,503
Specific Gravity 0.99
Formula Weight 136.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenyl-2-propanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.