missing translation for 'onlineSavingsMsg'
Learn More

1’N-Benzyl Biotin, TRC

Product Code. 17958914
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17958914 10 mg 10mg
30134312 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17958914 Supplier Toronto Research Chemicals Supplier No. TRCB23302510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76335-62-1
Formula Weight 334.1351
InChI Formula InChI=1S/C17H22N2O3S/c20-15(21)9-5-4-8-14-16-13(11-23-14)19(17(22)18-16)10-12-6-2-1-3-7-12/h1-3,6-7,13-14,16H,4-5,8-11H2,(H,18,22)(H,20,21)/t13-,14-,16-/m0/s1
IUPAC Name 5-[(3aR,6S,6aS)-3-benzyl-2-oxo-3a,4,6,6a-tetrahydro-1H-thieno[3,4-d]imidazol-6-yl]pentanoic acid
Molecular Formula C17H22N2O3S
Molecular Weight (g/mol) 334.43
SMILES OC(=O)CCCC[C@@H]1SC[C@H]2[C@@H]1NC(=O)N2Cc3ccccc3
Synonym (3aS,4S,6aR)-Hexahydro-2-oxo-1-(phenylmethyl)-1H-thieno[3,4-d]imidazole-4-pentanoic Acid, 1H-Thieno[3,4-d]imidazole-4-pentanoic acid, hexahydro-2-oxo-1-(phenylmethyl)-, [3aS-(3aα,4β,6aα)]-, Thieno[3,4-d]imidazoline-4-valeric acid, 1-benzyltetrahydro-2-oxo- (7CI), 5-[(3aS,4S,6aR)-3-Benzyl-2-oxohexahydrothieno[3,4-d]imidazol-4-yl]pentanoic Acid
Chemical Name or Material 1'N-Benzyl Biotin

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.