missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 97%, Thermo Scientific™

Product Code. p-3484816
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11380627

Brand: Thermo Scientific Maybridge CC45839CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-06-0
Molecular Formula C15H20BNO2
Molecular Weight (g/mol) 257.14
MDL Number MFCD08690266
InChI Key CBEYYYCGOCYJIK-UHFFFAOYSA-N
Synonym 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1h-indole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methylindol-4-boronic acid pinacol ester, 1-methylindol-4-boronic acid, pinacol ester, 1-methyl-1h-indol-4-yl boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-1-methylindol-4-yl-1,3,2-dioxaborolane, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-indole
PubChem CID 18525785
SMILES CN1C=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Formula Weight 257.14
Percent Purity 97%
Chemical Name or Material 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.