missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(3-nitrophenyl)piperazine, 97%, Thermo Scientific™

Product Code. p-3175562
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10183714 1 g 1g
10036724 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10183714 Supplier Thermo Scientific Maybridge Supplier No. CC35925DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 148546-97-8
Molecular Formula C11H15N3O2
Molecular Weight (g/mol) 221.26
MDL Number MFCD08435858
InChI Key IIRKKCDXJIXWHI-UHFFFAOYSA-N
Synonym 1-methyl-4-3-nitrophenyl piperazine, 4-methyl-1-3-nitrophenyl piperazine, piperazine,1-methyl-4-3-nitrophenyl, zlchem 196, pubchem18685, acmc-209d0u, 1-3-nitrophenyl 4-methylpiperazine, 1-3-nitrophenyl-4-methylpiperazine, 1-methyl-4-3-nitrophenyl-piperazine, 1-methyl-4-3-nitro-phenyl-piperazine
PubChem CID 12152161
IUPAC Name 1-methyl-4-(3-nitrophenyl)piperazine
SMILES CN1CCN(CC1)C1=CC(=CC=C1)[N+]([O-])=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.26
Percent Purity 97%
Chemical Name or Material 1-Methyl-4-(3-nitrophenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.