missing translation for 'onlineSavingsMsg'
Learn More

1-Iododecane, 98%, stab. with copper

Product Code. p-7023099
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
This item is not returnable. View return policy

Product Code. 11484160

Brand: Thermo Scientific Alfa Aesar A16173.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2050-77-3 | C10H21I | 268.18 g/mol

1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-77-3
Molecular Formula C10H21I
Molecular Weight (g/mol) 268.18
MDL Number MFCD00001085
InChI Key SKIDNYUZJPMKFC-UHFFFAOYSA-N
PubChem CID 16314
IUPAC Name 1-iododecane
SMILES CCCCCCCCCCI

Specifications

Melting Point -16°C
Density 1.257
Boiling Point 264°C
Flash Point 96°C (204°F)
Linear Formula CH3(CH2)9I
Refractive Index 1.485
Quantity 25 g
Beilstein 1735228
Sensitivity Light sensitive
Synonym decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942
Solubility Information Immiscible with water.
Formula Weight 268.18
Percent Purity 98%
Chemical Name or Material 1-Iododecane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.