Learn More
Fisherbrand™ 1 Hole Natural Rubber Stopper
Product Code. 11537622 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Bottom:
79 mm
99 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bottom unitSize
11537622 79 mm Each
11557622 99 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11537622
Only null left
Add to basket
Add to basket
Prevents liquid chemical leaks or escape from container.
  • Small, tapered plug used to seal the openings of test tubes, flasks and other laboratory glassware
  • Preferable for applications that require tighter seal or greater degree of chemical resistance
  • Prevent contamination of samples by protecting contents of laboratory glassware from air
  • Allows experimenters to shake or mix solutions without spilling them
Specifications
Hardness 46/55 IRHD
Height (Metric) 54mm
Quantity 1/Ea.
Color Red
Diameter (Metric) Bottom 79 mm
Material Natural Rubber
Product Type Tapered Stopper (1 hole)