missing translation for 'onlineSavingsMsg'
Learn More

1-Heptene, 98%

Product Code. p-3523272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10358243 25 mL Glass bottle 25mL
10453595 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10358243 Supplier Thermo Scientific Acros Supplier No. 120390250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-76-7 | C7H14 | 98.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-76-7
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00009531
InChI Key ZGEGCLOFRBLKSE-UHFFFAOYSA-N
Synonym 1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum
PubChem CID 11610
IUPAC Name hept-1-ene
SMILES CCCCCC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -119.0°C
Color Colorless
Density 0.6970g/mL
Boiling Point 94.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH=CH2
Refractive Index 1.3995
Quantity 25 mL
Beilstein 01, 219
Specific Gravity 0.697
Formula Weight 98.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Heptene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Heptene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: 1-Heptene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.