missing translation for 'onlineSavingsMsg'
Learn More

1-(Furan-2-ylcarbonyl)piperazine, Mikromol™

Product Code. 30050950
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050950 100 mg 100mg
30052145 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050950 Supplier Mikromol Supplier No. MM0085.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-(Furan-2-ylcarbonyl)piperazine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(furan-2-ylcarbonyl)piperazine
CAS 40172-95-0
Analyte or Component Names 1-(furan-2-ylcarbonyl)piperazine
Shipping Condition Room Temperature
Molecular Formula C9 H12 N2 O2
Certifications/Compliance ISO 17025
Synonym Prazosin Hydrochloride Imp. D (EP), Prazosin Imp. D (EP), Terazosin Hydrochloride Dihydrate Imp. L (EP), Terazosin Imp. L (EP), 1-(Furan-2-ylcarbonyl)piperazine, Prazosin Hydrochloride Impurity D, Terazosin Hydrochloride Dihydrate Impurity L, Prazosin Impurity D, Terazosin Impurity L
InChI Formula InChI=1S/C9H12N2O2/c12-9(8-2-1-7-13-8)11-5-3-10-4-6-11/h1-2,7,10H,3-6H2
SMILES O=C(N1CCNCC1)c2occc2
IUPAC Name furan-2-yl(piperazin-1-yl)methanone
Molecular Weight (g/mol) 180.2
Quantity 100 mg
Formula Weight 180.0899 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.