missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Ethyl-2-iodobenzene, 98%

Product Code. 11431858
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11431858

Brand: Alfa Aesar™ A14509.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
1-Ethyl-2-iodobenzene is used as pharmaceutical intermediate and as a reagent. It is used to prepare ethyl-2′-isopropylbiphenyl.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-40-1
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00040871
InChI Key ZEJZDNMOGNUIHL-UHFFFAOYSA-N
PubChem CID 140367
IUPAC Name 1-ethyl-2-iodobenzene
SMILES CCC1=CC=CC=C1I

Specifications

Boiling Point 221°C
Flash Point 101°C (213°F)
Quantity 50 g
Beilstein 2077313
Synonym 2-ethyliodobenzene, 1-iodo-2-ethylbenzene, benzene, 1-ethyl-2-iodo, 2-ethyl iodobenzene, pubchem3876, 1-ethyl-2-iodo-benzene, acmc-1brg2, #, benzene,1-ethyl-2-iodo, attercop-chm at134049
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 1-Ethyl-2-iodobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.