Learn More
1-Ethyl-2,3-dimethylimidazolium hexafluorophosphate, 98%, Thermo Scientific™
Product Code. 15465576
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 15465576
Brand: Thermo Scientific Alfa Aesar H60228.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 292140-86-4
Molecular Formula C7H13F6N2P
Molecular Weight (g/mol) 270.159
MDL Number MFCD08458503
InChI Key XLKYEJKYOZNSDQ-UHFFFAOYSA-N
Synonym 1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 3-ethyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, dsstox_cid_29120, dsstox_rid_83339, dsstox_gsid_49264, 1-ethyl-2,3-dimethylimidazol-1-ium hexafluorophosphate
PubChem CID 50878556
IUPAC Name 1-ethyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate
SMILES CCN1C=C[N+](=C1C)C.F[P-](F)(F)(F)(F)F
Specifications
Quantity 10 g
Formula Weight 270.16
Percent Purity 98%
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium hexafluorophosphate