missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-propanol, tech. 75% (remainder mainly 2-chloro-1-propanol)

Product Code. p-7026606
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445327 25 g 25g
11455327 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445327 Supplier Thermo Scientific Alfa Aesar Supplier No. B22686.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-00-4 | C3H7ClO | 94.538 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-00-4
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.538
MDL Number MFCD00004530
InChI Key YYTSGNJTASLUOY-UHFFFAOYSA-N
Synonym 1-chloro-2-propanol, 2-propanol, 1-chloro, chloroisopropyl alcohol, propene chlorohydrin, 1-chloroisopropyl alcohol, sec-propylene chlorohydrin, 1-chloro-2-hydroxypropane, .alpha.-propylene chlorohydrin, ccris 4766, 3-chloro-2-propanol
PubChem CID 31370
ChEBI CHEBI:76260
IUPAC Name 1-chloropropan-2-ol
SMILES CC(CCl)O

Specifications

Density 1.111
Boiling Point 126°C to 127°C
Flash Point 51°C (123°F)
Refractive Index 1.437
Quantity 25 g
UN Number UN2929
Beilstein 773653
Merck Index 14,7850
Formula Weight 94.54
Percent Purity 75%
Grade Technical
Assay remainder mainly 2-chloro-1-propanol
Chemical Name or Material 1-Chloro-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.