Learn More
1-Bromotridecane-d27, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Bromotridecane-D27 |
| CAS | 349553-91-9 |
| Molecular Formula | C13 D27 Br |
| Quantity | 0.25 g |
| Synonym | Tridecane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13-d27, 13-bromo- (9 CI), 13-Bromotridecane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13-d27, 13-Bromotridecane-d27, 1-Bromotridecane-d27, 1-Bromotridecane-D27, Tridecyl-d27 bromide |
| InChI Formula | InChI=1 S/C13H27Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h2-13H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,13-heptacosadeuteriotridecane |
| Molecular Weight (g/mol) | 290.4239 |
| Formula Weight | 289.2991 g/mol |
| Show More |