missing translation for 'onlineSavingsMsg'
Learn More

1-Azakenpaullone, MedChemExpress

Product Code. 16470356
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16470356

Brand: Medchem Express HY59090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 1-Azakenpaullone (1-Akp) is a highly selective and ATP-competitive inhibitor of glycogen synthase kinase-3 β (GSK-3β), with an IC50 value of 18 nM.

1-Azakenpaullone (1-Akp) is a highly selective and ATP-competitive inhibitor of glycogen synthase kinase-3 β (GSK-3β), with an IC50 value of 18 nM.

Specifications

Chemical Name or Material 1-Azakenpaullone
CAS 676596-65-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H10BrN3O
Quantity 2 mg
Synonym 1-Akp
Solubility Information DMSO : 16.67 mg/mL (50.80 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CN=C12)CC3=C2NC4=C3C=C(Br)C=C4
Molecular Weight (g/mol) 328.16
Formula Weight 328.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.