Learn More
1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid, 97%, Thermo Scientific™
Product Code. 11317096
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317096 5 g 5g
11327096 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11317096 Supplier Thermo Scientific Alfa Aesar Supplier No. L07019.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1779-10-8
Molecular Formula C11H6Br2O3
Molecular Weight (g/mol) 345.97
MDL Number MFCD00046367
InChI Key WNMKUIQCIRAXBN-UHFFFAOYSA-N
Synonym 4,7-dibromo-3-hydroxy-2-naphthoic acid, 1,6-dibromo-2-hydroxynaphthalene-3-carboxylic acid, 2-naphthalenecarboxylic acid, 4,7-dibromo-3-hydroxy, 1,6-dibromo-2-naphthol-3-carboxylic acid, 2-naphthoic acid,7-dibromo-3-hydroxy, 2-naphthalenecarboxylic acid,7-dibromo-3-hydroxy, acmc-1btqt, 2-naphthoic acid, 4,7-dibromo-3-hydroxy, labotest-bb lt00455402, 1,6-dibromo-2-hydroxynaphthalene-3-carb
PubChem CID 74502
IUPAC Name 4,7-dibromo-3-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=C(O)C(Br)=C2C=CC(Br)=CC2=C1
Specifications
Melting Point 249°C to 253°C
Odor Odorless
Quantity 5 g
Beilstein 2378969
Formula Weight 345.98
Percent Purity 97%
Chemical Name or Material 1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid