Learn More
(1,5-Cyclooctadiene)(hexafluoroacetylacetonato)iridium(I), TRC
Specifications
Specifications
| Chemical Name or Material | (1,5-Cyclooctadiene)(hexafluoroacetylacetonato)iridium(I) |
| CAS | 34801-95-1 |
| Molecular Formula | C8 H12 . C7 H9 F6 Ir O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C8H12.C5H2F6O2.2CH4.Ir/c1-2-4-6-8-7-5-3-1;6-4(7,8)2(12)1-3(13)5(9,10)11;;;/h1-2,7-8H,3-6H2;1,12H;2*1H4;/q;;;;+1/p-1/b2-1-,8-7-;;;; |
| SMILES | FC(F)(F)C1=[O][Ir](=[O]C(=C1)C(F)(F)F)([CH4])[CH4].C2C\C=C/CC\C=C/2 |
| IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene;2,2-di(methyl)-4,6-bis(trifluoromethyl)-1λ3,3λ3-dioxa-2λ5-iridacyclohexa-1,3,5-triene |
| Molecular Weight (g/mol) | 539.534 |
| Formula Weight | 540.107 |