missing translation for 'onlineSavingsMsg'
Learn More

1,4-Pentanediol, TRC

Product Code. 17929444
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17929444 25 g 25g
17979734 5 g 5g
17958664 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17929444 Supplier Toronto Research Chemicals Supplier No. TRCP27340025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-95-9
Molecular Formula C5 H12 O2
Molecular Weight (g/mol) 104.148
Synonym 1,4-Dihydroxy-1-methylbutane, 1,4-Dihydroxypentane, 2,5-Pentanediol, 4-Hydroxy-1-pentanol
IUPAC Name pentane-1,4-diol
SMILES CC(O)CCCO

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H12O2/c1-5(7)3-2-4-6/h5-7H,2-4H2,1H3
Formula Weight 104.0837
Chemical Name or Material 1,4-Pentanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.