Learn More
1-(4-Chloromethyl-2-thiazoyl)guanidine Hydrochloride Salt, TRC
Product Code. 30179878
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179878 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30179878 Supplier Toronto Research Chemicals Supplier No. TRCC3695451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69014-12-6
Molecular Formula C5 H7 Cl N4 S . Cl H
Molecular Weight (g/mol) 227.11
IUPAC Name 1-[4-(chloromethyl)-1,3-thiazol-2-yl]guanidine;hydrochloride
SMILES Cl.NC(=N)Nc1nc(CCl)cs1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C5H7ClN4S.ClH/c6-1-3-2-11-5(9-3)10-4(7)8;/h2H,1H2,(H4,7,8,9,10);1H
Formula Weight 225.9847
Chemical Name or Material 1-(4-Chloromethyl-2-thiazoyl)guanidine Hydrochloride Salt