missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylpyrazole, TRC

Product Code. 30387905
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387905 10 g
30371883 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387905 Supplier Toronto Research Chemicals Supplier No. TRCD47892510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-48-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
Synonym 2,5-Dimethylpyrazole, NSC 190569
IUPAC Name 1,3-dimethylpyrazole
SMILES Cc1ccn(C)n1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H8N2/c1-5-3-4-7(2)6-5/h3-4H,1-2H3
Formula Weight 96.0687
Chemical Name or Material 1,3-Dimethylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.