missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-n-butyl-2-thiobarbituric acid, 98%, Thermo Scientific Chemicals

Product Code. p-7027723
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11480588

Brand: Thermo Scientific Alfa Aesar B24707.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54443-89-9 | C12H20N2O2S | 256.364 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54443-89-9
Molecular Formula C12H20N2O2S
Molecular Weight (g/mol) 256.364
MDL Number MFCD00674068
InChI Key JNFBSDIDVPDZHB-UHFFFAOYSA-N
Synonym 1,3-dibutyl-2-thiobarbituric acid, 1,3-di-n-butyl-2-thiobarbituric acid, 4,6 1h,5h-pyrimidinedione, 1,3-dibutyldihydro-2-thioxo, 1,3-dibutyl-2-thioxodihydropyrimidine-4,6 1h,5h-dione, acmc-1asrh, 1,3-dibutyl-2-thioxohexahydropyrimidine-4,6-dione, 1,3-dibutyl-2-thioxo-1,3,5-trihydropyrimidine-4,6-dione
PubChem CID 108592
IUPAC Name 1,3-dibutyl-2-sulfanylidene-1,3-diazinane-4,6-dione
SMILES CCCCN1C(=O)CC(=O)N(C1=S)CCCC

Specifications

Melting Point 61°C to 63°C
Quantity 1 g
Formula Weight 256.37
Percent Purity 98%
Chemical Name or Material 1,3-Di-n-butyl-2-thiobarbituric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.