Learn More
1,3-Cyclooctadiene, 97%
Product Code. 11358475
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11358475
Brand: Thermo Scientific Alfa Aesar L10516.14
Request Bulk or Custom Format
CAS: 1700-10-3 | C8H12 | 108.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1700-10-3
Molecular Formula C8H12
Molecular Weight (g/mol) 108.18
MDL Number MFCD00001751
InChI Key RRKODOZNUZCUBN-CCAGOZQPSA-N
Synonym 1,3-cyclooctadiene, cyclooctadiene, 1z,3z-cycloocta-1,3-diene, cis,cis-1,3-cyclooctadiene, cycloocta-1,3-diene, 1,3-cyclooctadiene, z,z, cyclooctadienes, 1.3-cyclooctadiene, 1e,3z-1,3-cyclooctadiene, 1,3-cod
PubChem CID 299882
IUPAC Name (1Z,3Z)-cycloocta-1,3-diene
SMILES C1CC\C=C/C=C\C1
Specifications
Melting Point -53°C
Density 0.869
Boiling Point 142°C to 144°C
Flash Point 24°C (75°F)
Odor Characteristic
Refractive Index 1.493
Quantity 25 g
UN Number UN2520
Beilstein 1901033
Formula Weight 108.18
Percent Purity 97%
Chemical Name or Material 1,3-Cyclooctadiene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3-Cyclooctadiene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Cyclooctadiene