missing translation for 'onlineSavingsMsg'
Learn More

[1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene] Dichloro(o-isopropoxyphenylmethylene)ruthenium, TRC

Product Code. 17974435
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17974435 500 mg 500mg
17942755 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17974435 Supplier Toronto Research Chemicals Supplier No. TRCB588655500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301224-40-8
Molecular Formula C31 H38 Cl2 N2 O Ru
Molecular Weight (g/mol) 626.62
Synonym (SP-5-41)-[1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro[[2-(1-methylethoxy-κO)phenyl]methylene-κC]ruthenium, (1,3-Dimesityl-2-imidazolidinylidene)(o-isopropoxyphenylmethylene)ruthenium Dichloride, Grubbs-Hoveyda 2nd Generation Catalyst, Grubbs-Hoveyda Catalyst, Hoveyda II Catalyst, Hoveyda Catalyst, Hoveyda Second Generation Catalyst, Hoveyda-Grubbs 2nd Generation Catalyst, Hoveyda-Grubbs Catalyst, Hoveyda-Grubbs Catalyst 2nd. Generation, Hoveyda-Grubbs Second Generation Catalyst, Second Generation Hoveyda Grubbs Catalyst, Second-generation Hoveyda Catalyst
IUPAC Name [1,3-bis(2,4,6-trimethylphenyl)imidazolidin-2-ylidene]-dichloro-[(2-isopropoxyphenyl)methylene]ruthenium
SMILES CC(C)Oc1ccccc1C=[Ru](=C2N(CCN2c3c(C)cc(C)cc3C)c4c(C)cc(C)cc4C)(Cl)Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H26N2.C10H12O.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-8(2)11-10-7-5-4-6-9(10)3;;;/h9-12H,7-8H2,1-6H3;3-8H,1-2H3;2*1H;/q;;;;+2/p-2
Formula Weight 626.1405
Chemical Name or Material [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene] Dichloro(o-isopropoxyphenylmethylene)ruthenium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.