Learn More
1,2-Dichloroethanol Acetate, TRC
Product Code. 30386706
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386706 1 g 1g
30386962 2.5 g 2.5g
30380606 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30386706 Supplier Toronto Research Chemicals Supplier No. TRCD4346151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10140-87-1
Molecular Formula C4H6Cl2O2
Molecular Weight (g/mol) 157
Synonym 1,2-Dichloroethyl Acetate
IUPAC Name 1,2-dichloroethyl acetate
SMILES CC(=O)OC(Cl)CCl
Specifications
Quantity 1 g
InChI Formula InChI=1S/C4H6Cl2O2/c1-3(7)8-4(6)2-5/h4H,2H2,1H3
Formula Weight 155.9745
Chemical Name or Material 1,2-Dichloroethanol Acetate