Learn More
1,2-Cyclohexanedicarboxylic acid, bis-isononyl ester 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30326429
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326429 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326429 Supplier Dr. Ehrenstorfer Supplier No. DREA11824600AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2-Cyclohexanedicarboxylic acid, bis-isononyl ester
CAS 166412-78-8
Analyte or Component Names 1,2-Cyclohexanedicarboxylic acid, bis-isononyl ester
Shipping Condition Room Temperature
Molecular Formula C26 H48 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Bis(isononyl)cyclohexane 1,2-Dicarboxylate, Diisononyl Cyclohexane-1,2-dicarboxylate, Diisononyl Hexahydrophthalate, 1,2-Diisononyl-1,2-Cyclohexanedicarboxylic Acid Ester, DINCH, Hexamoll DINCH
InChI Formula InChI=1S/C26H48O4/c1-21(2)15-9-5-7-13-19-29-25(27)23-17-11-12-18-24(23)26(28)30-20-14-8-6-10-16-22(3)4/h21-24H,5-20H2,1-4H3
SMILES O=C(C1C(C(OCCCCCCC(C)C)=O)CCCC1)OCCCCCCC(C)C
IUPAC Name bis(7-methyloctyl) cyclohexane-1,2-dicarboxylate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 424.67
Quantity 1 mL
Formula Weight 424.36 g/mol