Learn More
1-(2-Bromophenylsulfonyl)-3-methylpiperidine, 97%, Thermo Scientific™
Product Code. p-7040434
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445556 250 mg 250mg
15436688 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15445556 Supplier Thermo Scientific Alfa Aesar Supplier No. H60206.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Este artículo no se puede devolver. Vea la política de devoluciones
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1-(2-Bromophenylsulfonyl)-3-methylpiperidine is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 1184609-55-9
Molecular Formula C12H16BrNO2S
Molecular Weight (g/mol) 318.229
MDL Number MFCD12650340
InChI Key DHHOXSFLPKIJME-UHFFFAOYSA-N
Synonym 1-2-bromobenzenesulfonyl-3-methylpiperidine, 1-2-bromophenylsulfonyl-3-methylpiperidine
PubChem CID 51169806
IUPAC Name 1-(2-bromophenyl)sulfonyl-3-methylpiperidine
SMILES CC1CCCN(C1)S(=O)(=O)C2=CC=CC=C2Br
Especificaciones
Quantity 250 mg
Formula Weight 318.23
Percent Purity 97%
Chemical Name or Material 1-(2-Bromophenylsulfonyl)-3-methylpiperidine
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.