missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-isopropylidene-D-mannitol

Product Code. 11398625
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398625 5 g 5g
11308635 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398625 Supplier Thermo Scientific Alfa Aesar Supplier No. L12127.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1707-77-3 | C12H22O6 | 262.302 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1707-77-3
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.302
MDL Number MFCD00003211
InChI Key ODYBCPSCYHAGHA-ZYUZMQFOSA-N
Synonym 1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213
PubChem CID 96011
IUPAC Name (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
SMILES CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C

Specifications

Melting Point 120°C to 122°C
Quantity 5 g
Beilstein 83765
Optical Rotation +6° (c=5 in Chloroform)
Formula Weight 262.31
Chemical Name or Material 1,2:5,6-Di-O-isopropylidene-D-mannitol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.