missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrakis(Bromomethyl)Benzene, TRC

Product Code. 30133928
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133928 100 mg 100mg
30136474 500 mg 500mg
30005035 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133928 Supplier Toronto Research Chemicals Supplier No. TRCT298568100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15442-91-8
Formula Weight 445.7516
InChI Formula InChI=1S/C10H10Br4/c11-3-7-1-8(4-12)10(6-14)2-9(7)5-13/h1-2H,3-6H2
IUPAC Name 1,2,4,5-tetrakis(bromomethyl)benzene
Molecular Formula C10 H10 Br4
Molecular Weight (g/mol) 449.8
SMILES BrCc1cc(CBr)c(CBr)cc1CBr
Synonym Tetrabromodurene
Chemical Name or Material 1,2,4,5-Tetrakis(bromomethyl)benzene

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.