Learn More
1-[(1RS)-2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride, Mikromol™
Description
1-[(1RS)-2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 1-[(1RS)-2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride |
| CAS | 130198-05-9 |
| Analyte or Component Names | 1-[(1RS)-2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H23 N O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Venlafaxine Hydrochloride Imp. C (EP), Venlafaxine Imp. C (EP), 1-[(1RS)-2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride, Venlafaxine Hydrochloride Imp. C (EP) as Hydrochloride, Venlafaxine Hydrochloride Impurity C as Hydrochloride, Venlafaxine Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C15H23NO2.ClH/c1-18-13-7-5-12(6-8-13)14(11-16)15(17)9-3-2-4-10-15;/h5-8,14,17H,2-4,9-11,16H2,1H3;1H |
| SMILES | Cl.COc1ccc(cc1)C(CN)C2(O)CCCCC2 |
| IUPAC Name | 1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride |
| Show More |