Learn More
1-(1H-Benzimidazol-2-yl)-3-methyl-1H-pyrazol-5-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(1H-Benzimidazol-2-yl)-3-methyl-1H-pyrazol-5-ol |
| CAS | 516455-57-5 |
| Molecular Formula | C11H10N4O |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C11H10N4O/c1-7-6-10(16)15(14-7)11-12-8-4-2-3-5-9(8)13-11/h2-6,14H,1H3,(H,12,13) |
| SMILES | CC1=CC(=O)N(N1)c2nc3ccccc3[nH]2 |
| IUPAC Name | 2-(1H-benzimidazol-2-yl)-5-methyl-1H-pyrazol-3-one |
| Molecular Weight (g/mol) | 214.22 |
| Formula Weight | 214.0855 |