Learn More
1,1-Dimethylethyl (R)-[2-[(methylsulfonyl)oxy]-1-phenylethyl]Carbamate, TRC
Specifications
Specifications
| CAS | 102089-75-8 |
| Molecular Formula | C14H21NO5S |
| Quantity | 100 mg |
| Synonym | (R)-2-((tert-Butoxycarbonyl)amino)-2-phenylethyl Methanesulfonate |
| InChI Formula | InChI=1S/C14H21NO5S/c1-14(2,3)20-13(16)15-12(10-19-21(4,17)18)11-8-6-5-7-9-11/h5-9,12H,10H2,1-4H3,(H,15,16) |
| SMILES | CC(C)(C)OC(=O)NC(COS(=O)(=O)C)c1ccccc1 |
| IUPAC Name | [2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylethyl] methanesulfonate |
| Molecular Weight (g/mol) | 315.39 |
| Formula Weight | 315.114 |
| Chemical Name or Material | 1,1-Dimethylethyl (R)-[2-[(methylsulfonyl)oxy]-1-phenylethyl]Carbamate |