Learn More
1,1-Dimethyl 2-Ethenylcyclopropane-1,1-dicarboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1-Dimethyl 2-Ethenylcyclopropane-1,1-dicarboxylate |
| CAS | 17447-60-8 |
| Molecular Formula | C9H12O4 |
| Quantity | 500 mg |
| Synonym | Dimethyl 2-Vinylcyclopropane-1,1-dicarboxylate, Dimethyl 3-Vinylcyclopropane-1,1-dicarboxylate |
| InChI Formula | InChI=1S/C9H12O4/c1-4-6-5-9(6,7(10)12-2)8(11)13-3/h4,6H,1,5H2,2-3H3 |
| SMILES | COC(=O)C1(CC1C=C)C(=O)OC |
| IUPAC Name | dimethyl 2-ethenylcyclopropane-1,1-dicarboxylate |
| Molecular Weight (g/mol) | 184.19 |
| Formula Weight | 184.0736 |