missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A Hemagglutinin H1 Antibody (InA139) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-73160JF585
This item is not returnable.
View return policy
Description
Influenza A Hemagglutinin H1 Monoclonal antibody specifically detects Influenza A Hemagglutinin H1 in Virus samples. It is validated for ELISA,Western Blot,Sandwich ELISA
Specifications
| Influenza A Hemagglutinin H1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| HA, Hemagglutinin, Hemagglutinin HA | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/NewCaledonia/20/99 (strain H1N1). | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Western Blot, Sandwich ELISA | |
| InA139 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction