Learn More
JAM-A Antibody (H202-106) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NB100-65340JF669
Product Code. 30174091
  • 724.00 EUR / 0.25mL
Description
JAM-A Monoclonal antibody specifically detects JAM-A in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
JAM-A
Monoclonal
Janelia Fluor 669
CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1
MTE1/MTE2 stromal cell lines
0.25 mL
Primary
Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
H202-106
50mM Sodium Borate
Rat
Protein G purified
RUO
50848
Store at 4°C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions