Learn More
JAM-A Antibody (H202-106) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-65340JF669
This item is not returnable.
View return policy
Description
JAM-A Monoclonal antibody specifically detects JAM-A in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Frozen)
Specifications
| JAM-A | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
| MTE1/MTE2 stromal cell lines | |
| 0.25 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| H202-106 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| 50848 | |
| Store at 4°C in the dark. | |
| IgG1 |