missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zeta Opioid Receptor Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97488MFV450
This item is not returnable.
View return policy
Description
Zeta Opioid Receptor Polyclonal antibody specifically detects Zeta Opioid Receptor in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Zeta Opioid Receptor | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| 7-60, 7-60 protein, OGFr, opioid growth factor receptor, Protein 7-60, Zeta-type opioid receptor | |
| Produced in rabbits immunized with E. coli-derived Human Zeta Opioid Receptor fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction | |
| 11054 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction