Learn More
TGF-beta 1 Antibody (Genzyme patent anti-TGF-beta) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28452JF525
Product Code. 30172200
  • 431.00 EUR / 0.10mL
Description
TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TGF-beta 1
Monoclonal
Janelia Fluor 525
CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
TGFb1
0.1 mL
Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction
7040
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
Genzyme patent anti-TGF-beta
50mM Sodium Borate
Human
Protein A purified
RUO
Primary
Human
Purified