missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-C Antibody (Thrombogenics patent anti-PDGF-C) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28573JF585
This item is not returnable.
View return policy
Description
PDGF-C Monoclonal antibody specifically detects PDGF-C in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PDGF-C | |
| Monoclonal | |
| Janelia Fluor 585 | |
| FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E | |
| PDGFC/ VEGFE | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 56034 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Thrombogenics patent anti-PDGF-C | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction