missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicalin Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18435JF585
This item is not returnable.
View return policy
Description
Nicalin Polyclonal antibody specifically detects Nicalin in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Nicalin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 585 | |
| NET59, nicalin, nicalin homolog (zebrafish), nicalin homolog (zebrefish), Nicastrin-like protein | |
| Recognizes region between residues Between Between 375 and 425.. | |
| 0.1 mL | |
| Neuroscience | |
| 56926 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction